LIQUID NITROGEN TRAP
Supplier: Labconco
Acid Trap Insert^ Acid Trap Insert, Glacier white powder-coated steel construction, 2 valves may be opened (trap) or closed (by-pass). 3/4inch+ OD inlet and outlet connections, 20inch wire-reinforced PVC tubing, 3/4inch Inner diameter; and 2 clamps. Liquid nitrogen required