KIMBERLY-CLARK® KC200 KIMGUARD™ Sterilization Wrap, Sequential, KIMBERLY-CLARK PROFESSIONAL®
Supplier: Kimberly-Clark
KIMGUARD™
68030 68018 68036 68054
82007-860CS 342.95 USD
82007-860 82007-862 82007-866 37001-146
KIMBERLY-CLARK® KC200 KIMGUARD™ Sterilization Wrap, Sequential, KIMBERLY-CLARK PROFESSIONAL®
Sterilization Wraps
Available in six strengths, from KC100 to KC600, to offer precisely the right protection for your needs
KIMGUARD™ K200 Sterilization Wrap is the second of six strength levels. Printed with its name, size, and lot code.
Choosing is easy; the higher the number, the stronger the wrap. Sequential wrap. Used to package linen towels, packs, and gowns, light basin sets, and single instruments for sterilization. Low-lint fabric delivers barrier and strength properties that help prevent contamination after sterilization. Steam and EO compatible. Naturally latex-free. Nonsterile. Multiple sizes.
Ordering information: Case count indicates the number of sheets included.