Would you like to log in again?
Pressure Vessel HF100
Supplier: PERKIN ELMER
Reaction vessel for various applications. The 100 mL liner is made of PTFE-TFM (N3141014) and supported by the Vessel Jacket H100 Ceramics (N3141019). A PTFE-TFM Seal for Pressure Vessel M+H+XF (N3141015) with integrated relief valve and Safety Disk.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
PRESSURE VESSEL HF100 10136-572 N3141012 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty