GAS SPLIT KIT
Supplier: PERKIN ELMER
Gas Split Kit^ Includes: 150psi manifold regulator, 0-60psi gauge, 1/4in cap fitting, 1/4in nipple fitting, Junction clamp, And mounting bracket. Specifications: Max inlet pressure: 300psi, Max outlet pressure: 150psi, Inlet port: 1/4in ptf, Outlet port: 1/4in ptf, And Gauge port: 1/8in ptf.