Would you like to log in again?
Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.
SUPRA-CLEAN SPE COLUMNS FLPR (FLORISIL Z
Supplier: PERKIN ELMER
Column^ SPE Supra-Clean* FL-S (Florisil Pesticides Grade, Granular Shape), 2 g/25 mL, Florisil Pesticide (FL-S) Supra-Clean Silica-based Solid Phase Extraction (SPE) products are Can be useded for extracting polar compounds when analyzing pesticides.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SUPRA-CLEAN SPE COLUMNS FLPR (FLORISIL Z 10131-460 N9306447 Pack of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty