PETROLEUM COLORIMETER 115V
Supplier: KOEHLER INSTRUMENT COM INC
Petroleum Colorimeter^ Electrical Requirements: 115V 60Hz, Included Accessories: Glass Color Discs (2) Sample Container (3) Calibration Certificate, Dimensions dxwxh, in.(cm): 10.5x9x5 (25x27x18), Conforms to the specifications of: ASTM D1500, D6074.