SP Bel-Art Vial, Bottle, and Tube Rack, Bel-Art Products, a part of SP
Supplier: Bel-Art
SCIENCEWARE®
H18921-0000
71001-656CS 850.31 USD
71001-656
SP Bel-Art Vial, Bottle, and Tube Rack, Bel-Art Products, a part of SP
Racks Tube Racks
Compact, sturdy vial, bottle, and tube rack holds dropping bottles, vials, scintillation vials, and other containers.
  • Heavy-duty polypropylene construction
  • Chemically resistant
  • Autoclavable
  • Compact footprint
The 12-well rack has 4 wells with open fronts for complete viewing of contents. Each 33 mm diameter well holds containers from 28–32 mm. Stable, heavy-duty polypropylene construction is heat and chemical resistant and autoclavable at 121°C (250°F).