Methanol (7+3) in nitric acid 70% (v/v), VWR Chemicals BDH®

Supplier: VWR International

Danger

Synonyms: Hydroxy methaneMethyl hydroxideMethyl alcoholMethylolCarbinolWood alcohol

BDHVBDH7941-1
BDH7941-1CS 835.46 USD InStock
BDH7941-1
Methanol (7+3) in nitric acid 70% (v/v), VWR Chemicals BDH®
Methanol

Per liter formula:
700mLs ACS Methanol min 99.8% W/W
300mLs ACS Nitric Acid 68-70%W/W

Formula: CH₃OH
MW: 32,04 g/mol
Boiling Pt: 64.6 °C (1013 hPa)
Melting Pt: –98 °C
Density: 0.7918 g/cm³ (20 °C)
Flash Pt: 11 °C (closed cup)
Storage Temperature: ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3,II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://us.vwr.com/stibo/web/std.lang.all/33/12/733312.jpg

Order Now

Specification Test Results

Assay (Methanol) (v/v) 68.5 - 71.5 %
Assay (HNO3) (v/v) 28.5 - 31.5 %
Appearance/color Clear liquid
Specific gravity (15.5 °C) 0.980 - 1.020
Prepared to formulation on file Yes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR