Guanidinium hydrochloride 8 M, buffered, VWR Chemicals BDH®

Synonyms: Guanidinium chlorideGuanidine hydrochlorideGuanidine chlorideGuanadine hydrochloride

BDHVBDH7427-1
BDH7427-1CS 9290.21 USD
BDH7427-1
Guanidinium hydrochloride 8 M, buffered, VWR Chemicals BDH®
Guanidinium hydrochloride

In deionized water buffered with sodium hydroxide solution and TRIS hydrochloride.

Formula: NH₂C(=NH)NH₂·HCl
MW: 95.53 g/mol
MDL Number: MFCD00013026
CAS Number: 50-01-1
https://us.vwr.com/stibo/web/std.lang.all/74/24/2297424.jpg

Order Now

Specification Test Results

Appearance/color Clear liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR