Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methanol 20% (v/v), VWR Chemicals BDH®

Danger
Synonyms: Hydroxy methaneMethyl hydroxideMethyl alcoholMethylolCarbinolWood alcohol

BDH7905-4 BDHVBDH7905-4
BDH7905-4CS 689.12 USD InStock
CABDH7905-4 BDH7905-4
Methanol 20% (v/v), VWR Chemicals BDH®
Methanol
Per liter formula:
200mLs ACS Methanol min 99.8% W/W
800mLs Deionized Water


Formula: CH₃OH
MW: 32,04 g/mol
Boiling Pt: 64.6 °C (1013 hPa)
Melting Pt: –98 °C
Density: 0.7918 g/cm³ (20 °C)
Flash Pt: 11 °C (closed cup)
Storage Temperature: ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3,II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://us.vwr.com/stibo/web/std.lang.all/33/12/733312.jpg
Order Now
Specification Test Results
Assay (Methanol) (v/v) 18.0 - 22.0 %
Water (v/v) 78.0 - 82.0 %
Density 0.950 - 0.990
Appearance/color Clear liquid
Non-volatile residue 100 ppm max.

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty