Hydrogen peroxide 35% stabilized, VWR Chemicals BDH®

Danger

Synonyms: Dihydrogen dioxideHydrogen dioxideHydrogen oxide

BDHVBDH7814-3
BDH7814-3CS 207.29 USD
BDH7814-3
Hydrogen peroxide 35% stabilized, VWR Chemicals BDH®
Hydrogen peroxide

Flammable and corrosive.

Formula: H₂O₂
MW: 34.01 g/mol
Boiling Pt: ∼107 °C (1013 hPa)
Melting Pt: < 0 °C
Density: 1.11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011333
CAS Number: 7722-84-1
EINECS: 231-765-0
UN: 2014
ADR: 5.1,II

Order Now

Specification Test Results

Color (APHA) 25 max.
Shelf-life 2 years
pH 2.0 - 4.0
Assay 34.0 - 36.0 %
Stabilizer (organic phosphinate) Passes test
Specific gravity 1.130 - 1.140

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR