Sulfuric acid 0.25 N in aqueous solution, VWR Chemicals BDH®

Supplier: VWR International

BDHVBDH7678-4
BDH7678-4CS 333.68 USD
BDH7678-4
Sulfuric acid 0.25 N in aqueous solution, VWR Chemicals BDH®
Sulphuric acid

APHA. Made with high purity acid and deionized water. Suitable as a titrant.

Formula: H₂SO₄
MW: 98.08 g/mol
Density: 1.01 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 2796
ADR: 8,II

Order Now

Specification Test Results

Appearance/color Clear liquid
N.I.S.T. Traceable to SRM 723 Yes
Normality 0.2495 - 0.2505 N

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR