Sulfuric acid 5.0 N in aqueous solution, VWR Chemicals BDH®

Supplier: VWR International
Danger

Synonyms: Hydrogen sulfateOil of vitriolVitriol brown oil

BDHVBDH7648-1
BDH7648-1CS 1082.76 USD
BDH7648-1
Sulfuric acid 5.0 N in aqueous solution, VWR Chemicals BDH®
Sulphuric acid

Made with high purity acid and deionized water. Suitable as a titrant.

Formula: H₂SO₄
MW: 98.08 g/mol
Boiling Pt: 330 °C (1013 hPa)
Melting Pt: 10.38 °C
Density: 1.146 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 2796
ADR: 8,II
REACH: 01-2119458838-20

Order Now

Specification Test Results

Color (APHA) 10 max.
Normality 4.980 - 5.020 N
Appearance/color Clear liquid
N.I.S.T. Traceable to SRM 723 Yes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR