Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.

Potassium fluoride 50% (w/w) in aqueous solution, VWR Chemicals BDH®

Supplier: VWR International
Danger

BDH7455-1 BDHVBDH7455-1
BDH7455-1CS 1055.32 USD InStock
CABDH7455-1 BDH7455-1
Potassium fluoride 50% (w/w) in aqueous solution, VWR Chemicals BDH®
Potassium fluoride
Made with deionized water.


Formula: KF
MW: 58,1 g/mol
Boiling Pt: 1505 °C (1013 hPa)
Melting Pt: 860 °C
Density: 2.48 g/cm³ (20 °C)
Storage Temperature: ambient
MDL Number: MFCD00011398
CAS Number: 7789-23-3
EINECS: 232-151-5
UN: 3422
ADR: 6.1,III
Merck Index: 13,07714
Order Now
Specification Test Results
Appearance/color Clear liquid
Shelf-life 2 years
pH 8.0 - 8.2 of 1:6 dilution
Density 1.28 - 1.33

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty