Potassium permanganate 5% (w/v) in aqueous solution, VWR Chemicals BDH®

Supplier: VWR International

BDHVBDH7450-2
BDH7450-2CS 482.12 USD InStock
BDH7450-2
Potassium permanganate 5% (w/v) in aqueous solution, VWR Chemicals BDH®
Potassium permanganate

Made with potassium permanganate low mercury grade.

Formula: KMnO₄
MW: 158.03 g/mol
Melting Pt: Min. 240 °C
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011364
CAS Number: 7722-64-7
EINECS: 231-760-3
UN: 3082
ADR: 9,III
Merck Index: 12,07824

Order Now

Specification Test Results

Appearance/color Dark purple liquid
Potassium permanganate (w/v) 4.0 - 5.1 %
Shelf-life 1 year

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR