Boric acid 2% (w/v) in aqueous solution, VWR Chemicals BDH®

Synonyms: Boric acidBoron hydroxideBoron trihydroxideBoracic acidOrthoboric acid

BDHVBDH7386-4
BDH7386-4CS 556.89 USD
BDH7386-4
Boric acid 2% (w/v) in aqueous solution, VWR Chemicals BDH®
ortho-Boric acid

Made with deionized water.

Formula: B(OH)₃
MW: 61.83 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011337
CAS Number: 10043-35-3
EINECS: 233-139-2
https://us.vwr.com/stibo/web/std.lang.all/05/54/16640554.jpg

Order Now

Specification Test Results

Appearance/color Clear liquid
Shelf-life 1 year

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR