Mineral oil, white (petroleum), VWR Chemicals BDH®

Supplier: VWR International
Warning

Synonyms: Matrix oilMineral oilParaffin oil

BDHVBDH7338-4
BDH7338-4CS 645.67 USD
BDH7338-4
Mineral oil, white (petroleum), VWR Chemicals BDH®
White mineral oil (petroleum)
Boiling Pt: 300 °C (1013 hPa)
Density: 0.872 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00131611
CAS Number: 8042-47-5
EINECS: 232-455-8

Order Now

Specification Test Results

Appearance Clear liquid
Odour description Confirmed
Specific gravity (25°C/25°C) 0.852 -- 0.876
Viscosity (100°F) (ASTM-D2161) (SUS) 180.0 -- 190.0
Viscosity (40°C) (ASTM-D445) 34.9 -- 37.3 cSt

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR