Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nessler reagent in solution analytical reagent, VWR Chemicals BDH®

Danger
Synonyms: Potassium tetraiodomercurate solutionPotassium tetraiodomercurateMercury (II) potassium iodidePotassium tetraiodomercurate – Potassium hydroxide solutionPotassium mercuric Iodide

302V-20
BDH7332-2CS 1001.84 USD InStock
BDH7332-2
Nessler reagent in solution analytical reagent, VWR Chemicals BDH®
Nessler's reagent
Made with deionized water.


Formula: HgI₄K₂
MW: 786,4 g/mol
Boiling Pt: ∼100 °C (1013 hPa)
Density: 1.2 g/cm³ (20 °C)
MDL Number: MFCD00049656
CAS Number: 7783-33-7
EINECS: 000-000-0
UN: 2922
ADR: 8,II
Order Now
Specification Test Results
Appearance/color Clear yellow liquid
Shelf-life 1 year

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty