Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Manganese(II) sulfate monohydrate solution 364 g/L (38%), VWR Chemicals BDH®

Supplier: VWR International
Warning
Synonyms: Manganous sulfate monohydrateManganous sulphate monohydrateManganese sulfate monohydrateManganese sulphate monohydrate

BDH7329-1 BDHVBDH7329-1 BDHVBDH7329-1
BDH7329-1CS 753.28 USD InStock
CABDH7329-1 BDH7329-1 BDH7329-1
Manganese(II) sulfate monohydrate solution 364 g/L (38%), VWR Chemicals BDH®
Manganese (II) sulphate monohydrate
APHA and ASTM for DO (dissolved oxygen).


Formula: MnSO₄·1H₂O
MW: 169,02 g/mol
Melting Pt: 700 °C
Density: 2.95 g/cm³ (25 °C)
Storage Temperature: 2 - 8°C
MDL Number: MFCD00149159
CAS Number: 10034-96-5
EINECS: 232-089-9
UN: 3077
ADR: 9,III
Merck Index: 13,05763
Order Now
Specification Test Results
Appearance/color Clear pink liquid
Specific gravity (20°C) 1.268 - 1.272

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty