EDTA disodium salt 0.0575 M in aqueous solution, VWR Chemicals BDH®

Supplier: VWR International

Synonyms: Edetic acid disodium saltDisodium dihydrogen ethylenediaminetetraacetateEthylenediaminetetra-acetic acid disodium saltIdranal IIIKestranal® 2SEthylenedinitrilotetra-acetic acid disodium salt

BDHVBDH7315-1
BDH7315-1CS 1042.84 USD
BDH7315-1
EDTA disodium salt 0.0575 M in aqueous solution, VWR Chemicals BDH®
EDTA disodium salt

Suitable as a titrant. Made with deionized water.

Formula: C₁₀H₁₄N₂Na₂O₈
MW: 336.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00070672
CAS Number: 139-33-3
EINECS: 205-358-3

Order Now

Specification Test Results

Appearance/color Clear liquid
N.I.S.T. traceable to SRM 915 Yes
Molarity 0.0570 -0.0580 M
Shelf-life 2 years

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR