o-Tolidine solution 0,01% (w/v), VWR Chemicals BDH®

Supplier: VWR International

Synonyms: 4,4'-Bi-o-toluidine

BDHVBDH7241-1
BDH7241-1CS 524.56 USD InStock
BDH7241-1
o-Tolidine solution 0,01% (w/v), VWR Chemicals BDH®
o-Tolidine

Made with deionized water, hydrochloric acid and o-tolidine.

Formula: [-C₆H₃(CH₃)-₄-NH₂]₂
MW: 212,29 g/mol
Boiling Pt: ~ 361 °C (1013 hPa)
Melting Pt: 129 to 131 °C
Density: ~ 1.234 g/cm³ (20 °C)
Storage Temperature: ambient
MDL Number: MFCD00014773
CAS Number: 119-93-7
EINECS: 204-358-0
UN: 3077
ADR: 9,III
Merck Index: 12,09651

Order Now

Specification Test Results

Prepared to formulation on file Yes
Appearance/color Clear liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR