Potassium permanganate 0.1 N in aqueous solution, VWR Chemicals BDH®

Supplier: VWR International

BDHVBDH7213-4 BDHVBDH7213-1 BDHVBDH7213-7
BDH7213-4CS 294.57 USD InStock
BDH7213-4 BDH7213-1 BDH7213-7
Potassium permanganate 0.1 N in aqueous solution, VWR Chemicals BDH®
Potassium permanganate

Suitable to be used as a titrant

Formula: KMnO₄
MW: 158.03 g/mol
Melting Pt: Min. 240 °C
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011364
CAS Number: 7722-64-7
EINECS: 231-760-3
UN: 3082
ADR: 9,III
Merck Index: 12,07824

Order Now

Specification Test Results

Appearance/color Dark purple clear liquid
N.I.S.T. Traceable to SRM 8040 Yes
Normality 0.0995-0.1005 N
Shelf-life 1 year

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR