Perchloric acid 0.1 N in acetic acid, VWR Chemicals BDH®

Supplier: VWR International
Danger

301V-03 301V-20 301V-06
BDH7210-4CS 1247.23 USD
BDH7210-4 BDH7210-2 BDH7210-1
Perchloric acid 0.1 N in acetic acid, VWR Chemicals BDH®
Perchloric acid

Made with high purity acetic acid, suitable as a titrant.

Formula: HClO₄
MW: 100.46 g/mol
Boiling Pt: 39 °C (75 hPa)
Melting Pt: -101 °C
Storage Temperature: Ambient
MDL Number: MFCD00011325
CAS Number: 7601-90-3
EINECS: 231-512-4
UN: 2920
ADR: 8,II
https://us.vwr.com/stibo/web/std.lang.all/06/22/16640622.jpg

Order Now

Specification Test Results

Appearance/color Clear liquid
N.I.S.T. Traceable Yes
Normality 0.0995 - 0.1005 N
Shelf-life 1 year

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR