Would you like to log in again?
Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.
Jackson Safety® G60 Level 2 Nitrile Coated Cut Glove, KIMBERLY-CLARK PROFESSIONAL®
Supplier: KIMBERLY CLARK CORP
JACKSON SAFETY®
98232 98231 98230 98233 98234
89140-866CS 772.01 USD InStock
89140-866 89140-864 89140-862 89140-868 89140-870
Jackson Safety® G60 Level 2 Nitrile Coated Cut Glove, KIMBERLY-CLARK PROFESSIONAL®
Gloves
Add cut resistance to your workers' gloves without significantly adding to your costs with the JACKSON SAFETY® G60 Level 2 Nitrile Coated Cut Gloves
A great choice for working environments that pose a risk of minor cuts and abrasions. Stretch KEVLAR® liner provides cut resistance and a comfortable fit. Gloves also feature an ANSI/EN 388 Level 2 Cut Resistance Abrasion resistant nitrile coating. Nitrile coating enhances grip in light oil applications and the dark color hides dirt for longer wear. Excellent dexterity for improved productivity. ACTIFRESH® antimicrobial treatment inhibits bacteria growth, thereby extending wear and improving cost-in-use. Suitable for intermittent heat contact up to 212˚F. Glove must not come into contact with a naked flame. Ideally suited for metal fabrication, glass handling, automotive and aerospace assembly, stamping, and assembly.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty