Blue Nitrile Gloves, Kimberly-Clark
Supplier: Kimberly-Clark
KIMBERLY-CLARK®
53103 53100 53101 53104 53102
89003-042CS 538.33 USD
89003-042 89003-040 89003-034 89003-038 89003-036
Blue Nitrile Gloves, Kimberly-Clark
Gloves
Nitrile gloves offer excellent tactile sensitivity and dexterity as well as superior fit and comfort.
  • Textured fingertips enhance wet and dry grip
  • Ambidextrous and powder-free
  • Single use
Beaded cuff for added strength and easier donning.