4,5-Dimethoxy-2-nitrobenzyl carbonochloridate 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4,5-Dimethoxy-2-nitrobenzyl chloroformate

L14167-03 L14167-03
AAL14167-03EA 153.99 USD
CAAAL14167-03 AAL14167-03
4,5-Dimethoxy-2-nitrobenzyl carbonochloridate 97%
4,5-Dimethoxy-2-nitrobenzyl carbonochloridate
Formula: C₁₀H₁₀ClNO₆
MW: 275.65 g/mol
Melting Pt: ca 120 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00143507
CAS Number: 42855-00-5
UN: 3261
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR