1-(2-Cyanoethyl)piperidine ≥98%

Warning

Synonyms: Piperidinopropionitrile

L01469-30 L01469-18
AAL01469-30EA 148.96 USD
AAL01469-30 AAL01469-18
1-(2-Cyanoethyl)piperidine ≥98%
1-(2-Cyanoethyl)piperidine
Formula: C₈H₁₄N₂
MW: 138.21 g/mol
Boiling Pt: 110…111 °C/16 mm
Density: 0.933
Flash Pt: 102 °C(215 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006511
CAS Number: 3088-41-3
EINECS: 221-417-6
UN: 3276
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR