DAPT

Supplier: Alfa Aesar

Synonyms: (3,5-Difluorophenylacetyl)-Ala-Phg-OtBu(3,5-Difluorophenylacetyl)-L-alanyl-L-2-phenylglycine tert-Butyl Ester

J65864-MA J65864-MA
AAJ65864-MAEA 234.05 USD
AAJ65864-MA CAAAJ65864-MA
DAPT
DAPT

Lyophilized


Applications: A cell-permeable dipeptide that suppresses A production by blocking -secretase

Formula: C₂₃H₂₆F₂N₂O₄
MW: 432.47 g/mol
Storage Temperature: Freezer
MDL Number: MFCD04974585
CAS Number: 208255-80-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR