2',3'-Dideoxyguanosine ≥90%

Synonyms: 2-Amino-9-((2R,5S)-5-(hydroxymethyl)tetrahydrofuran-2-yl)-1H-purin-6(9H)-one

J65810-MB J65810-MB J65810-MF J65810-MC J65810-MC J65810-MF
AAJ65810-MCEA 306.95 USD
CAAAJ65810-MC CAAAJ65810-MB AAJ65810-MC AAJ65810-MB AAJ65810-MF CAAAJ65810-MF
2',3'-Dideoxyguanosine ≥90%
2',3'-Dideoxyguanosine

Powder

Formula: C₁₀H₁₃N₅O₃
MW: 251.24
Storage Temperature: Refrigerator
MDL Number: MFCD00057074
CAS Number: 85326-06-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR