2'-Deoxycytidine-5'-triphosphoric acid disodium salt (dCTP-Na2)

Synonyms: 2'-Deoxycytidine-5'-triphosphate disodium salt2′-Deoxycytidine-5′-Triphosphate Trisodium Salt

J65686-MD J65686-MD
AAJ65686-MDEA 309.5 USD
AAJ65686-MD CAAAJ65686-MD
2'-Deoxycytidine-5'-triphosphoric acid disodium salt (dCTP-Na2)
2'-Deoxycytidine-5'-triphosphoric acid disodium salt (dCTP-Na2)

Powder

Formula: C₉H₁₄N₃Na₂O₁₃P₃
MW: 511.12
Storage Temperature: Refrigerator
MDL Number: MFCD00084683
CAS Number: 102783-51-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR