R5C3, 5-Methyl-N-(2-phenoxybenzoyl)-1-[(phenylmethoxy)carbonyl]-D/L-tryptophan

J65639-MA J65639-MA
AAJ65639-MAEA 287.84 USD
AAJ65639-MA CAAAJ65639-MA
R5C3, 5-Methyl-N-(2-phenoxybenzoyl)-1-[(phenylmethoxy)carbonyl]-D/L-tryptophan
R5C3, 5-Methyl-N-(2-phenoxybenzoyl)-1-[(phenylmethoxy)carbonyl]-D/L-tryptophan

A MDM2 inhibitor and 2-phenoxybenzoyl-tryptophan derivative. Reported as a novel probe for high throughput fluorescence polarization binding assay


Powder


Reported as a novel probe for high throughput fluorescence polarization binding assay

Formula: C₃₃H₂₈N₂O₆
MW: 548.58 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08705330
CAS Number: 753504-14-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR