2,4,6-Trimethyl-N-(2-(trifluoromethyl)phenyl)benzenesulfonamide

Supplier: Thermo Scientific Chemicals

Warning

J65391-MF J65391-MA J65391-MF J65391-MA
AAJ65391-MAEA 123 USD
AAJ65391-MA CAAAJ65391-MF AAJ65391-MF CAAAJ65391-MA
2,4,6-Trimethyl-N-(2-(trifluoromethyl)phenyl)benzenesulfonamide
2,4,6-Trimethyl-N-(2-(trifluoromethyl)phenyl)benzenesulphonamide

An inactive analog of m-3M3FBS. Used as a negative control.


Powder

Formula: C₁₆H₁₆F₃NO₂S
MW: 343.36 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00784362
CAS Number: 313981-55-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR