FBPase-1 inhibitor
|
Warning
|
Synonyms:
2,5-Dichloro-N-(5-chlorobenzo[d]oxazol-2-yl)benzenesulfonamide
, 2,5-Dichloro-N-(5-chlorobenzo[d]oxazol-2-yl)benzenesulphonamide
J65298-MF
J65298-MA
J65298-MA
J65298-MF
AAJ65298-MAEA
202.61
USD
AAJ65298-MA
CAAAJ65298-MA
AAJ65298-MF
CAAAJ65298-MF
FBPase-1 inhibitor
FBPase-1 inhibitor
A cell-permeable benzoxazolo-sulfonamide compound that inhibits hFBPase-1
- Light Sensitive
Solid
Soluble in DMSO
Formula:
C₁₃H₇Cl₃N₂O₃S MW: 377.60 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22581250 CAS Number: 883973-99-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...