Apoptosis inhibitor

Synonyms: M50054

J65036-MA J65036-MA
AAJ65036-MAEA 142.76 USD
CAAAJ65036-MA AAJ65036-MA
Apoptosis inhibitor
Apoptosis inhibitor

A cell-permeable inhibitor of apoptosis induction. Inhibits caspase-3 activation but does not directly inhibit caspase-3 even at 1 mg/ml.


  • Light Sensitive


Solid


Soluble in DMSO or ethanol

Formula: C₁₃H₁₆O₄
MW: 236.00 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00192291
CAS Number: 54135-60-3
EINECS: 258-989-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR