ALFAJ64979
J64979-MA J64979-MA
AAJ64979-MAEA 180.54 USD
AAJ64979-MA CAAAJ64979-MA
ALFAJ64979
Powder
  • Light Sensitive
  • Hygroscopic
Applications: A cell-permeable pyrimidine compound that specifically and reversibly inhibits Rac1 GDP/GTP exchange activity