HNHA

Warning

J64917-MA J64917-MA
AAJ64917-MAEA 209.41 USD
CAAAJ64917-MA AAJ64917-MA
HNHA
HNHA

A cell-permeable hydroxyamide-based SAHA analog that potently inhibits histone deacetylase (HDAC) activity


  • Light Sensitive


Crystalline solid

Formula: C₁₇H₂₁NO₂S
MW: 303.42 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10568146
CAS Number: 926908-04-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR