EBPC

Danger

J64858-MA J64858-MA
AAJ64858-MAEA 118.44 USD
AAJ64858-MA CAAAJ64858-MA
EBPC
EBPC

A potent aldose reductase inhibitor


Solid

Formula: C₁₄H₁₅NO₄
MW: 261.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00179167
CAS Number: 57056-57-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR