Phenylethyl 3-methylcaffeate

J64722-MC J64722-MF J64722-MF J64722-MC
AAJ64722-MCEA 314.28 USD
AAJ64722-MC AAJ64722-MF CAAAJ64722-MF CAAAJ64722-MC
Phenylethyl 3-methylcaffeate
Phenylethyl 3-methylcaffeate

Induces apoptosis and inhibits carcinogenesis in animal models of colon cancer


Solid

Formula: C₁₈H₁₈O₄
MW: 298.33 g/mol
Melting Pt: 80…81 °C
Storage Temperature: Ambient
MDL Number: MFCD18385026
CAS Number: 71835-85-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR