2'-Bromo-2'-deoxyuridine 98%

Supplier: Thermo Scientific Chemicals

J64704-ME J64704-ME
AAJ64704-MEEA 165.37 USD
CAAAJ64704-ME AAJ64704-ME
2'-Bromo-2'-deoxyuridine 98%
2'-Bromo-2'-deoxyuridine

Powder

Formula: C₉H₁₁BrN₂O₅
MW: 307.10 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06658226
CAS Number: 72218-68-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR