2',3'-Dideoxyadenosine 98%

Supplier: Thermo Scientific Chemicals

Synonyms: ((2S,5R)-5-(6-Amino-9H-purin-9-yl)tetrahydrofuran-2-yl)methanol

J64573-MB J64573-MF J64573-MF J64573-MB J64573-MC J64573-MC
AAJ64573-MCEA 495.51 USD
CAAAJ64573-MB CAAAJ64573-MC CAAAJ64573-MF AAJ64573-MC AAJ64573-MB AAJ64573-MF
2',3'-Dideoxyadenosine 98%
2',3'-Dideoxyadenosine

Powder

Formula: C₁₀H₁₃N₅O₂
MW: 235.24 g/mol
Melting Pt: 181…184 °C
Storage Temperature: Ambient
MDL Number: MFCD00010534
EINECS: 223-853-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR