GSK-3β inhibitor VIII
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1-(4-Methoxybenzyl)-3-(5-nitrothiazol-2-yl)urea
, N-(4-Methoxybenzyl)-N′-(5-nitro-1,3-thiazol-2-yl)urea
, AR-A014418
J64554-MA
J64554-MA
AAJ64554-MAEA
197.79
USD
AAJ64554-MA
CAAAJ64554-MA
GSK-3β inhibitor VIII
GSK-3beta inhibitor VIII
A cell-permeable, potent, and selective inhibitor of GSK-3beta
- Light Sensitive
Solid
Formula:
C₁₂H₁₂N₄O₄S MW: 308.31 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08277040 CAS Number: 487021-52-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...