2-Chloro-5-nitro-N-(4-pyridyl)benzamide

Warning

Synonyms: 2-Chloro-5-nitro-N-(pyridin-4-yl)benzamide

J64455-MF J64455-MF J64455-MA J64455-MA
AAJ64455-MAEA 150.97 USD
CAAAJ64455-MF AAJ64455-MA AAJ64455-MF CAAAJ64455-MA
2-Chloro-5-nitro-N-(4-pyridyl)benzamide
2-Chloro-5-nitro-N-(4-pyridyl)benzamide

A cell-permeable chloro-nitro-benzamido compound that acts as a potent, specific, irreversible, and high-affinity antagonist of PPAR


Solid

Formula: C₁₂H₈ClN₃O₃
MW: 277.7 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00121849
CAS Number: 313516-66-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR