Trypsin 1:250, porcine pancreas

Supplier: Alfa Aesar
J63993-18 J63993-18 J63993-09 J63993-09
AAJ63993-18EA 256.98 USD InStock
CAAAJ63993-18 AAJ63993-18 AAJ63993-09 CAAAJ63993-09
Trypsin 1:250, porcine pancreas
Enzymes
Powder

CAS No.: 9002-07-7
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White to off-White
MDL No.: MFCD01775455

Applications: A pancreatic serine protease
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR