TRIS-Tricine-SDS Kit (10X), Thermo Scientific

Supplier: Thermo Scientific
J63948-K2 J63948-K2 J63948-AP J63948-AP
AAJ63948-APEA 89.94 USD
AAJ63948-AP CAAAJ63948-AP AAJ63948-K2 CAAAJ63948-K2
TRIS-Tricine-SDS Kit (10X), Thermo Scientific
Buffers Electrophoresis Buffers Transfer Buffers
Contains one each of Tris anode buffer and Tris-Tricine-SDS cathode buffer for PAGE.

  • Liquid
  • Store at Ambient Temperatures
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR