8-Bromoguanosine

Supplier: Thermo Scientific Chemicals

Synonyms: 2-Amino-8-bromo-9-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-1H-purin-6(9H)-one

J63940-06 J63940-06
AAJ63940-06EA 136.96 USD
AAJ63940-06 CAAAJ63940-06
8-Bromoguanosine
8-Bromoguanosine

Powder

Formula: C₁₀H₁₂BrN₅O₅
MW: 362.14 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00037985
CAS Number: 4016-63-1
EINECS: 223-677-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR