Carbonate, 0.5M buffer soln., pH 9.5

Supplier: Alfa Aesar
J63658-AK J63658-AP J63658-AK J63658-AP
AAJ63658-AKEA 80.23 USD
AAJ63658-AK AAJ63658-AP CAAAJ63658-AK CAAAJ63658-AP
Carbonate, 0.5M buffer soln., pH 9.5
Buffers Biological Buffers
Liquid

CAS No.: 497-19-8
Physical Form: Liquid
Odor: Odorless
MDL No.: MFCD00003494
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR