HEPES, 1.0M buffer soln., pH 8.0

Supplier: Alfa Aesar
J63578-AP J63578-AK J63578-AP J63578-AK
AAJ63578-APEA 242.28 USD
AAJ63578-AP AAJ63578-AK CAAAJ63578-AP CAAAJ63578-AK
HEPES, 1.0M buffer soln., pH 8.0
Buffers Biological Buffers
Liquid

CAS No.: 7365-45-9
Physical Form: Liquid
MDL No.: MFCD00006158
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR