Agarose Gel Loading Dye 6X for Electrophoresis Bromophenol-free, Glycerol Based, Thermo Scientific

Supplier: Thermo Scientific
J63429-AC J63429-AD J63429-AC J63429-AD
AAJ63429-ACEA 62.66 USD
CAAAJ63429-AC CAAAJ63429-AD AAJ63429-AC AAJ63429-AD
Agarose Gel Loading Dye 6X for Electrophoresis Bromophenol-free, Glycerol Based, Thermo Scientific
Contains 0.25% xylene cyanol FF and 30% glycerol.

  • Free from Bromophenol blue
  • Store at Ambient temperatures

Liquid
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR