Glipizide

Supplier: Thermo Scientific Chemicals

Synonyms: N-[4-(3-Cyclohexylureidosulfonyl)phenethyl]-5-methyl-2-pyrazinecarboxamide

J63398-03 J63398-06 J63398-06 J63398-03
AAJ63398-03EA 101.23 USD
CAAAJ63398-03 AAJ63398-03 AAJ63398-06 CAAAJ63398-06
Glipizide
Glipizide

An ATP-dependent potassium channel blocker


Powder

Formula: C₂₁H₂₇N₅O₄S
MW: 445.54 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00072159
CAS Number: 29094-61-9
EINECS: 249-427-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR