Clindamycin phosphate

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Clindamycin 2-Dihydrogen Phosphate

J63387-MC J63387-MC
AAJ63387-MCEA 284.05 USD
CAAAJ63387-MC AAJ63387-MC
Clindamycin phosphate
Clindamycin phosphate

Antibiotic U-28508E, effective against Gram-positive bacteria


Antibiotic U-28508E, effective against gram-positive bacteria

Formula: C₁₈H₃₄ClN₂O₈PS
MW: 504.96 g/mol
Melting Pt: 114 °C
Storage Temperature: Ambient
MDL Number: MFCD07793328
CAS Number: 24729-96-2
EINECS: 246-433-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR